accessibility menu, dialog, popup

UserName

CAS RN 7446-08-4

OSeO

CAS RN 7446-08-4

IUPAC Name: (oxo-λ⁴-selanylidene)oxidane
Synonyms: selenium dioxide
Molecular Weight (g/mol): 110.97
Molecular Formula: O2Se
InChi Key: JPJALAQPGMAKDF-UHFFFAOYSA-N
SMILES: O=[Se]=O
Physical Form 
  • (2)
  • (2)
  • (6)
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Grade 
  • (1)
  • (1)
  • (6)
Percent Purity 
  • (2)
  • (4)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (6)
  • (4)
  • (2)
  • (2)
  • (6)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (6)
  • (2)
  • (4)
  • (2)
  • (1)