CAS RN 7440-03-1
IUPAC Name:
niobium
Synonyms:
niobium columbium columbio niobio Niob niobium atom
Molecular Weight (g/mol):
92.91
Molecular Formula:
Nb
InChi Key:
GUCVJGMIXFAOAE-UHFFFAOYSA-N
SMILES:
[Nb]
1
–
2
of
2
results
1
Niobium Standard For ICP, 1000μg/mL in H2O/0.4% HF, Assurance Grade, SPEX CertiPrep™
CAS: 7440-03-1 Molecular Formula: Nb Molecular Weight (g/mol): 92.91 MDL Number: MFCD00011126 InChI Key: GUCVJGMIXFAOAE-UHFFFAOYSA-N PubChem CID: 23936 ChEBI: CHEBI:33344 IUPAC Name: niobium SMILES: [Nb]
2
Niobium Standard For ICP, 10,000μg/mL in H2O/0.4% HF, SPEX CertiPrep™
CAS: 7440-03-1 Molecular Formula: Nb Molecular Weight (g/mol): 92.91 MDL Number: MFCD00011126 InChI Key: GUCVJGMIXFAOAE-UHFFFAOYSA-N PubChem CID: 23936 ChEBI: CHEBI:33344 IUPAC Name: niobium SMILES: [Nb]