accessibility menu, dialog, popup

UserName

CH3H3C

CAS RN 74-84-0

IUPAC Name: ethane
Synonyms: ethane dimethyl ethanes aethan ethane (substance) bimethyl
Molecular Weight (g/mol): 30.07
Molecular Formula: C2H6
InChi Key: OTMSDBZUPAUEDD-UHFFFAOYSA-N
SMILES: CC
Solvent or Matrix 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results