accessibility menu, dialog, popup

UserName

CAS RN 7333-51-9

BrP

CAS RN 7333-51-9

IUPAC Name: cyclohexyltriphenylphosphanium bromide
Synonyms: cyclohexyltriphenylphosphanium bromide
Molecular Weight (g/mol): 425.35
Molecular Formula: C24H26BrP
InChi Key: QRAKRDZMIONMRZ-UHFFFAOYSA-M
SMILES: [Br-].C1CCC(CC1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results