accessibility menu, dialog, popup

UserName

CAS RN 7242-92-4

(R)(R)(S)H2N

CAS RN 7242-92-4

IUPAC Name:
Synonyms:
Molecular Weight (g/mol): 404.4
Molecular Formula: C6H14O8Os
InChi Key: CGFZRSJVZXGPJR-UHFFFAOYSA-N
SMILES: O=[Os]=O.CC(O)O.CC(O)=O.CC(O)=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results