accessibility menu, dialog, popup

UserName

CAS RN 71989-18-9

H3CCH3H3COOHOONHOO

CAS RN 71989-18-9

IUPAC Name: 5-(tert-butoxy)-4-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-5-oxopentanoic acid
Synonyms: 5-(tert-butoxy)-4-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-5-oxopentanoic acid
Molecular Weight (g/mol): 425.48
Molecular Formula: C24H27NO6
InChi Key: GOPWHXPXSPIIQZ-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)C(CCC(O)=O)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results