CAS RN 71868-10-5
IUPAC Name:
2-methyl-1-[4-(methylsulfanyl)phenyl]-2-(morpholin-4-yl)propan-1-one
Synonyms:
methylthiophenyl morpholino isobutanone
Molecular Weight (g/mol):
279.4
Molecular Formula:
C15H21NO2S
InChi Key:
LWRBVKNFOYUCNP-UHFFFAOYSA-N
SMILES:
CSC1=CC=C(C=C1)C(=O)C(C)(C)N1CCOCC1
1
–
2
of
2
results
1
2-Methyl-1-(4-(methylthio)phenyl)-2-morpholinopropan-1-one, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2-Methyl-1-(4-(methylthio)phenyl)-2-morpholinopropan-1-one, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.