accessibility menu, dialog, popup

UserName

CAS RN 71759-88-1

CH3NNI

CAS RN 71759-88-1

IUPAC Name: 5-iodo-1-methyl-1H-imidazole
Synonyms: 5-iodo-1-methylimidazole
Molecular Weight (g/mol): 208
Molecular Formula: C4H5IN2
InChi Key: DPUVICGLBCCTDM-UHFFFAOYSA-N
SMILES: CN1C=NC=C1I

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results