CAS RN 708-79-2
CAS RN 708-79-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1-Methyluric Acid, TRC
CAS: 708-79-2 Molecular Formula: C6 H6 N4 O3 Molecular Weight (g/mol): 182.14 Synonym: 7,9-Dihydro-1-methyl-1H-purine-2,6,8(3H)-trione,1-Methyl-uric Acid; IUPAC Name: 1-methyl-7,9-dihydro-3H-purine-2,6,8-trione SMILES: CN1C(=O)NC2=C(NC(=O)N2)C1=O