accessibility menu, dialog, popup

UserName

CAS RN 70702-47-5

NOOHH2N

CAS RN 70702-47-5

IUPAC Name: 2-amino-3-(pyridin-3-yl)propanoic acid
Synonyms: 2-amino-3-(pyridin-3-yl)propanoic acid
Molecular Weight (g/mol): 166.18
Molecular Formula: C8H10N2O2
InChi Key: DFZVZEMNPGABKO-UHFFFAOYNA-N
SMILES: NC(CC1=CN=CC=C1)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)