accessibility menu, dialog, popup

UserName

CAS RN 704-38-1

OSS

CAS RN 704-38-1

IUPAC Name: bis(thiophen-2-yl)methanone
Synonyms: di-thiophen-2-yl-methanone
Molecular Weight (g/mol): 194.27
Molecular Formula: C9H6OS2
InChi Key: GUTQMBQKTSGBPQ-UHFFFAOYSA-N
SMILES: O=C(C1=CC=CS1)C1=CC=CS1
Molecular Weight (g/mol) 
  • (3)
  • (5)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Industry Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results