accessibility menu, dialog, popup

UserName

CAS RN 6970-19-0

CH3OOH3COH3COHO

CAS RN 6970-19-0

IUPAC Name: 3,4,5-triethoxybenzoic acid
Synonyms: 3,4,5-triethoxybenzoic acid
Molecular Weight (g/mol): 254.28
Molecular Formula: C13H18O5
InChi Key: YCENSLDYFDZUMO-UHFFFAOYSA-N
SMILES: CCOC1=CC(=CC(OCC)=C1OCC)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)