accessibility menu, dialog, popup

UserName

CAS RN 696-71-9

OH

CAS RN 696-71-9

IUPAC Name: cyclooctanol
Synonyms: cyclooctanol
Molecular Weight (g/mol): 128.22
Molecular Formula: C8H16O
InChi Key: FHADSMKORVFYOS-UHFFFAOYSA-N
SMILES: OC1CCCCCCC1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)