accessibility menu, dialog, popup

UserName

CAS RN 6954-77-4

ClOBrCl

CAS RN 6954-77-4

IUPAC Name: 1-(2-bromoethoxy)-2,4-dichlorobenzene
Synonyms: 1-(2-bromoethoxy)-2,4-dichlorobenzene
Molecular Weight (g/mol): 269.95
Molecular Formula: C8H7BrCl2O
InChi Key: OEYZGSRLAGSENP-UHFFFAOYSA-N
SMILES: ClC1=CC=C(OCCBr)C(Cl)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results