accessibility menu, dialog, popup

UserName

CAS RN 6948-33-0

IUPAC Name: (2E)-3-(3-bromo-4-hydroxy-5-methoxyphenyl)prop-2-enoate
Synonyms: (2E)-3-(3-bromo-4-hydroxy-5-methoxyphenyl)prop-2-enoate
Molecular Weight (g/mol): 272.08
Molecular Formula: C10H8BrO4
InChi Key: NGBVBJQUJJKRGF-NSCUHMNNSA-M
SMILES: COC1=CC(\C=C\C([O-])=O)=CC(Br)=C1O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results