CAS RN 69308-37-8
CAS RN 69308-37-8
IUPAC Name:
4-amino-3-(4-chlorophenyl)butanoic acid
Synonyms:
baclofen nu baclo novartis brand of baclofen lioresal intrathecal medtronic brand of baclofen NuBaclo baclofen awd baclofen alphapharm brand baclofene baclofeno
Molecular Weight (g/mol):
213.66
Molecular Formula:
C10H12ClNO2
InChi Key:
KPYSYYIEGFHWSV-UHFFFAOYNA-N
SMILES:
NCC(CC(O)=O)C1=CC=C(Cl)C=C1