accessibility menu, dialog, popup

UserName

CAS RN 690632-30-5

CH3OOSOOCl

CAS RN 690632-30-5

IUPAC Name: 4-(2-methoxyphenoxy)benzene-1-sulfonyl chloride
Synonyms: 4-(2-methoxyphenoxy)benzenesulfonyl chloride
Molecular Weight (g/mol): 298.74
Molecular Formula: C13H11ClO4S
InChi Key: DKNWPXFCLUKCQB-UHFFFAOYSA-N
SMILES: COC1=CC=CC=C1OC1=CC=C(C=C1)S(Cl)(=O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results