CAS RN 6724-53-4
CAS RN 6724-53-4
IUPAC Name:
(2Z)-but-2-enedioic acid; 2-(2,2-dicyclohexylethyl)piperidine
Synonyms:
maleic acid; perhexiline
Molecular Weight (g/mol):
393.57
Molecular Formula:
C23H39NO4
InChi Key:
JDZOTSLZMQDFLG-BTJKTKAUNA-N
SMILES:
OC(=O)\C=C/C(O)=O.C(C(C1CCCCC1)C1CCCCC1)C1CCCCN1
1
–
2
of
2
results
1
Tocris Bioscience™ Perhexiline maleate, >99%
CAS: 6724-53-4 Molecular Formula: C23H39NO4
2
STEMCELL Technologies Perhexiline Maleate
Carnitine palmitoyltransferase 1 (CPT1) and CPT2 inhibitor