CAS RN 6587-24-2
CAS RN 6587-24-2
IUPAC Name:
methyl 2-cyanobenzoate
Synonyms:
methyl 2-cyanobenzoate
Molecular Weight (g/mol):
161.16
Molecular Formula:
C9H7NO2
InChi Key:
RAMPDACRJWTXEV-UHFFFAOYSA-N
SMILES:
COC(=O)C1=CC=CC=C1C#N
1
–
3
of
3
results
1
Methyl 2-cyanobenzoate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Methyl 2-cyanobenzoate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Methyl 2-cyanobenzoate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.