CAS RN 65322-85-2
CAS RN 65322-85-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
3-(4-Isobutylphenyl)propanoic Acid, TRC
CAS: 65322-85-2 Molecular Formula: C13 H18 O2 Molecular Weight (g/mol): 206.28 Synonym: 3-[4-(2-Methylpropyl)phenyl]propanoic Acid,Ibuprofen Imp. F (EP) IUPAC Name: 3-[4-(2-methylpropyl)phenyl]propanoic acid SMILES: CC(C)Cc1ccc(CCC(=O)O)cc1
2
Ibuprofen EP Impurity F, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.