CAS RN 653-11-2
IUPAC Name:
(2,3,5,6-tetrafluorophenyl)hydrazine
Synonyms:
(2,3,5,6-tetrafluorophenyl)hydrazine
Molecular Weight (g/mol):
180.11
Molecular Formula:
C6H4F4N2
InChi Key:
TYMFVEOXNZYYTF-UHFFFAOYSA-N
SMILES:
NNC1=C(F)C(F)=CC(F)=C1F
1
–
1
of
1
results
1
2,3,5,6-Tetrafluorophenylhydrazine, 97%
CAS: 653-11-2 Molecular Formula: C6H4F4N2 Molecular Weight (g/mol): 180.11 MDL Number: MFCD00007575 InChI Key: TYMFVEOXNZYYTF-UHFFFAOYSA-N Synonym: 2,3,5,6-tetrafluorophenyl hydrazine,2,3,5,6-tetrafluorphenyl hydrazin,pubchem3300,acmc-20aoi4,tymfveoxnzyytf-uhfffaoysa,2,3,5,6-tetrafluoro-phenylhydrazine,1-2,3,5,6-tetrafluorophenyl hydrazine,1-hydrazino-2,3,5,6-tetrafluorobenzene,hydrazine, 2,3,5,6-tetrafluorophenyl,1-2,3,5,6-tetrafluorophenyl hydrazine # PubChem CID: 69552 IUPAC Name: (2,3,5,6-tetrafluorophenyl)hydrazine SMILES: NNC1=C(F)C(F)=CC(F)=C1F