accessibility menu, dialog, popup

UserName

CAS RN 651734-64-4

CH3OFNH2F

CAS RN 651734-64-4

IUPAC Name: 3-ethoxy-2,6-difluoroaniline
Synonyms: 3-ethoxy-2,6-difluoroaniline
Molecular Weight (g/mol): 173.16
Molecular Formula: C8H9F2NO
InChi Key: GHEWYZSXEPQBOO-UHFFFAOYSA-N
SMILES: CCOC1=C(F)C(N)=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results