CAS RN 65145-14-4
CAS RN 65145-14-4
IUPAC Name:
N,N-dibenzyl-4-bromoaniline
Synonyms:
N,N-dibenzyl-4-bromoaniline
Molecular Weight (g/mol):
352.28
Molecular Formula:
C20H18BrN
InChi Key:
RSFLPIGKWXARAL-UHFFFAOYSA-N
SMILES:
BrC1=CC=C(C=C1)N(CC1=CC=CC=C1)CC1=CC=CC=C1
1
–
1
of
1
results
1
N,N-Dibenzyl-4-bromoaniline, 95%, Thermo Scientific™
CAS: 65145-14-4 Molecular Formula: C20H18BrN Molecular Weight (g/mol): 352.28 MDL Number: MFCD09037473 InChI Key: RSFLPIGKWXARAL-UHFFFAOYSA-N Synonym: 4-bromo-n,n-dibenzylaniline,benzenemethanamine, n-4-bromophenyl-n-phenylmethyl,acmc-20ak9j,4-bromophenyldibenzylamine,n-4-bromophenyl dibenzylamine,n,n-dibenzyl-4-bromobenzenamine,4-n,n-dibenzylamino bromobenzene,n-4bromophenyl-n-phenylmethyl benzenemethanamine,benzenemethanamine,n-4-bromophenyl-n-phenylmethyl PubChem CID: 285423 IUPAC Name: N,N-dibenzyl-4-bromoaniline SMILES: BrC1=CC=C(C=C1)N(CC1=CC=CC=C1)CC1=CC=CC=C1