CAS RN 651-83-2
IUPAC Name:
2,3,5,6-tetrafluoro-4-(trifluoromethyl)aniline
Synonyms:
2,3,5,6-tetrafluoro-4-(trifluoromethyl)aniline
Molecular Weight (g/mol):
233.09
Molecular Formula:
C7H2F7N
InChi Key:
FJOACTZFMHZHSC-UHFFFAOYSA-N
SMILES:
NC1=C(F)C(F)=C(C(F)=C1F)C(F)(F)F
1
–
1
of
1
results
1
2,3,5,6-Tetrafluoro-4-(trifluoromethyl)aniline, 98%
CAS: 651-83-2 Molecular Formula: C7H2F7N Molecular Weight (g/mol): 233.089 MDL Number: MFCD00091518 InChI Key: FJOACTZFMHZHSC-UHFFFAOYSA-N Synonym: 2,3,5,6-tetrafluoro-4-trifluoromethyl aniline,2,3,5,6-tetrafluoro-4-aminobenzotrifluoride,4-amino-heptafluorotoluene,4-amino-2,3,5,6-tetrafluorobenzotrifluoride,4-aminotetrafluorobenzotrifluoride,4-aminoheptafluorotoluene,2,3,5,6-tetrafluoro-4-trifluoromethyl phenylamine,benzenamine, 2,3,5,6-tetrafluoro-4-trifluoromethyl,pubchem2817,yu-wasa auxiliary PubChem CID: 616257 IUPAC Name: 2,3,5,6-tetrafluoro-4-(trifluoromethyl)aniline SMILES: C1(=C(C(=C(C(=C1F)F)N)F)F)C(F)(F)F