accessibility menu, dialog, popup

UserName

CAS RN 6457-49-4

HONH2

CAS RN 6457-49-4

IUPAC Name: 4-(hydroxymethyl)piperidin-1-ium
Synonyms: 4-(hydroxymethyl)piperidin-1-ium
Molecular Weight (g/mol): 116.18
Molecular Formula: C6H14NO
InChi Key: XBXHCBLBYQEYTI-UHFFFAOYSA-O
SMILES: OCC1CC[NH2+]CC1
Molecular Weight (g/mol) 
  • (6)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (6)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (3)