accessibility menu, dialog, popup

UserName

CAS RN 64-77-7

IUPAC Name: 3-butyl-1-(4-methylbenzenesulfonyl)urea
Synonyms: tolbutamide dolipol orinase diagnostic drabet restinon willbutamide diabetamid tolylsulfonylbutylurea orezan tolbutamida
Molecular Weight (g/mol): 270.35
Molecular Formula: C12H18N2O3S
InChi Key: JLRGJRBPOGGCBT-UHFFFAOYSA-N
SMILES: CCCCNC(=O)NS(=O)(=O)C1=CC=C(C)C=C1
Molecular Weight (g/mol) 
  • (3)
  • (4)
Certifications/Compliance 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Industry Type 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results