accessibility menu, dialog, popup

UserName

CAS RN 62667-64-5

CH3OOCH3OOCH3OOEu

CAS RN 62667-64-5

IUPAC Name: tris(acetic acid) europium hydrate
Synonyms: tris(acetic acid) europium hydrate
Molecular Weight (g/mol): 350.14
Molecular Formula: C6H14EuO7
InChi Key: RWVNKGWAXJFQSB-UHFFFAOYSA-N
SMILES: O.[Eu].CC(O)=O.CC(O)=O.CC(O)=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)