accessibility menu, dialog, popup

UserName

CAS RN 626-43-7

NH2ClCl

CAS RN 626-43-7

IUPAC Name: 3,5-dichloroaniline
Synonyms: 3,5-dichloroaniline m-dichloroaniline
Molecular Weight (g/mol): 162.01
Molecular Formula: C6H5Cl2N
InChi Key: UQRLKWGPEVNVHT-UHFFFAOYSA-N
SMILES: NC1=CC(Cl)=CC(Cl)=C1
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)