accessibility menu, dialog, popup

UserName

OHBrBr

CAS RN 626-41-5

IUPAC Name: 3,5-dibromophenol
Synonyms: 3,5-dibromophenol
Molecular Weight (g/mol): 251.91
Molecular Formula: C6H4Br2O
InChi Key: PZFMWYNHJFZBPO-UHFFFAOYSA-N
SMILES: OC1=CC(Br)=CC(Br)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results