accessibility menu, dialog, popup

UserName

ClOCl

CAS RN 618-46-2

IUPAC Name: 3-chlorobenzoyl chloride
Synonyms: M-chlorobenzoylchloride
Molecular Weight (g/mol): 175.01
Molecular Formula: C7H4Cl2O
InChi Key: WHIHIKVIWVIIER-UHFFFAOYSA-N
SMILES: ClC(=O)C1=CC=CC(Cl)=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (4)
  • (3)
Percent Purity 
  • (3)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (3)
  • (3)
  • (4)
  • (4)