CAS RN 61774-58-1
CAS RN 61774-58-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Cis-Pinic Acid, TRC
CAS: 61774-58-1 Molecular Formula: C9H14O4 Molecular Weight (g/mol): 186.21 Synonym: (1R,3R)-rel-Cyclobutaneacetic acid, 3-carboxy-2,2-dimethyl-,cis-3-Carboxy-2,2-dimethyl-cyclobutaneacetic acid,cis-3-Carboxy-2,2-dimethylcyclobutaneacetic acid IUPAC Name: (1R,3R)-3-(carboxymethyl)-2,2-dimethylcyclobutane-1-carboxylic acid SMILES: OC([C@H]1C(C)(C)[C@@H](CC(O)=O)C1)=O