accessibility menu, dialog, popup

UserName

H3COH3CSOH3C

CAS RN 61295-41-8

IUPAC Name: 3-[(2-methylfuran-3-yl)sulfanyl]heptan-4-one
Synonyms: 3-[(2-methylfuran-3-yl)sulfanyl]heptan-4-one
Molecular Weight (g/mol): 226.33
Molecular Formula: C12H18O2S
InChi Key: GFRRQSASJZMMJC-UHFFFAOYNA-N
SMILES: CCCC(=O)C(CC)SC1=C(C)OC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results