accessibility menu, dialog, popup

UserName

CAS RN 612-28-2

H3CNHNOO

CAS RN 612-28-2

IUPAC Name: N-methyl-2-nitroaniline
Synonyms: N-methyl-O-nitroaniline
Molecular Weight (g/mol): 152.15
Molecular Formula: C7H8N2O2
InChi Key: KFBOUJZFFJDYTA-UHFFFAOYSA-N
SMILES: CNC1=CC=CC=C1[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results