accessibility menu, dialog, popup

UserName

CH3OHBr

CAS RN 60710-39-6

IUPAC Name: 3-bromo-4-methylphenol
Synonyms: 3-bromo-4-methylphenol
Molecular Weight (g/mol): 187.04
Molecular Formula: C7H7BrO
InChi Key: GMZKNRDHSHYMHG-UHFFFAOYSA-N
SMILES: CC1=C(Br)C=C(O)C=C1
Molecular Weight (g/mol) 
  • (1)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)