accessibility menu, dialog, popup

UserName

CAS RN 603-11-2

IUPAC Name: 3-nitrobenzene-1,2-dicarboxylic acid
Synonyms: 3-nitrophthalic acid
Molecular Weight (g/mol): 211.13
Molecular Formula: C8H5NO6
InChi Key: KFIRODWJCYBBHY-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CC(=C1C(O)=O)[N+]([O-])=O
Molecular Weight (g/mol) 
  • (4)
  • (6)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results