accessibility menu, dialog, popup

UserName

CAS RN 5990-32-9

OOOOOOOHOOOOOOHOH2OH2OZnZnZn

CAS RN 5990-32-9

IUPAC Name: trizinc(2+) bis(2-hydroxypropane-1,2,3-tricarboxylate) dihydrate
Synonyms: trizinc(2+) dicitrate dihydrate
Molecular Weight (g/mol): 610.37
Molecular Formula: C12H14O16Zn3
InChi Key: OXAGUGIXGVHDGD-UHFFFAOYSA-H
SMILES: O.O.[Zn++].[Zn++].[Zn++].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O.OC(CC([O-])=O)(CC([O-])=O)C([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)
Grade 
  • (2)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results