accessibility menu, dialog, popup

UserName

CAS RN 58846-77-8

H3CO(R)O(R)OH(S)OH(S)OH(R)HO

CAS RN 58846-77-8

IUPAC Name: (2R,3R,4S,5S,6R)-2-(decyloxy)-6-(hydroxymethyl)oxane-3,4,5-triol
Synonyms: decyl glucoside
Molecular Weight (g/mol): 320.43
Molecular Formula: C16H32O6
InChi Key: JDRSMPFHFNXQRB-IBEHDNSVSA-N
SMILES: CCCCCCCCCCO[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)