CAS RN 57696-89-6
CAS RN 57696-89-6
IUPAC Name:
3,5,5-trimethylcyclohexane-1,2-dione
Synonyms:
3,5,5-trimethylcyclohexane-1,2-dione
Molecular Weight (g/mol):
154.21
Molecular Formula:
C9H14O2
InChi Key:
WEUZZYGESXCTCU-UHFFFAOYNA-N
SMILES:
CC1CC(C)(C)CC(=O)C1=O
1
–
1
of
1
results
1
3,5,5-Trimethylcyclohexane-1,2-dione, 99%, Thermo Scientific™
CAS: 57696-89-6 Molecular Formula: C9H14O2 Molecular Weight (g/mol): 154.209 MDL Number: MFCD02181023 InChI Key: WEUZZYGESXCTCU-UHFFFAOYSA-N Synonym: 3,5,5-trimethyl-1,2-cyclohexanedione,1,2-cyclohexanedione, 3,5,5-trimethyl,3,5,5-trimethyl-1.2-cyclohexanedione,acmc-1aooj,ksc274g5l,3,5,5-trimethyl-1,2 cyclohexanedione,3,5,5-trimethyl cyclohexane-1,2-dione,3,5,5-trimethylcyclohexane-1,2-dione 5g PubChem CID: 93772 IUPAC Name: 3,5,5-trimethylcyclohexane-1,2-dione SMILES: CC1CC(CC(=O)C1=O)(C)C