accessibility menu, dialog, popup

UserName

KOHOHOHHOOOHOHO

CAS RN 576-42-1

IUPAC Name: 2,3,4,5-tetrahydroxyhexanedioic acid potassium
Synonyms: D-saccharate potassium
Molecular Weight (g/mol): 249.24
Molecular Formula: C6H10KO8
InChi Key: AHPUWJRVCCLSPB-UHFFFAOYNA-N
SMILES: [K].OC(C(O)C(O)C(O)=O)C(O)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results