accessibility menu, dialog, popup

UserName

BrNOO

CAS RN 574-98-1

IUPAC Name: 2-(2-bromoethyl)-2,3-dihydro-1H-isoindole-1,3-dione
Synonyms: N-(2-bromoethyl)phthalimide
Molecular Weight (g/mol): 254.08
Molecular Formula: C10H8BrNO2
InChi Key: CHZXTOCAICMPQR-UHFFFAOYSA-N
SMILES: BrCCN1C(=O)C2=CC=CC=C2C1=O
Molecular Weight (g/mol) 
  • (6)
  • (3)
Quantity 
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results