CAS RN 57384-79-9
CAS RN 57384-79-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
1-(2-oxopropyl)-2,4-dihydro-1H-3,1-benzoxazine-2,4-dione, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
1-(2-oxopropyl)-2,4-dihydro-1H-3,1-benzoxazine-2,4-dione, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
1-(2-oxopropyl)-2,4-dihydro-1H-3,1-benzoxazine-2,4-dione, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.