CAS RN 57079-01-3
IUPAC Name:
11-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)undecanoic acid
Synonyms:
11-(2,5-dioxopyrrol-1-yl)undecanoic acid
Molecular Weight (g/mol):
281.35
Molecular Formula:
C15H23NO4
InChi Key:
UVZTZBRGZXIBLZ-UHFFFAOYSA-N
SMILES:
OC(=O)CCCCCCCCCCN1C(=O)C=CC1=O
1
–
2
of
2
results
1
11-Maleimidoundecanoic acid
CAS: 57079-01-3 Molecular Formula: C15H23NO4 Molecular Weight (g/mol): 281.352 MDL Number: MFCD00941279 InChI Key: UVZTZBRGZXIBLZ-UHFFFAOYSA-N Synonym: 11-maleimidoundecanoic acid,11-2,5-dioxo-2,5-dihydro-1h-pyrrol-1-yl undecanoic acid,unii-lg78ey7hvb,n-10-carboxydecyl maleimide,11-2,5-dioxopyrrol-1-yl undecanoic acid,lg78ey7hvb,maleimide-ch2 10-cooh,1h-pyrrole-1-undecanoic acid, 2,5-dihydro-2,5-dioxo,kmua,pubchem11837 PubChem CID: 4618600 IUPAC Name: 11-(2,5-dioxopyrrol-1-yl)undecanoic acid SMILES: C1=CC(=O)N(C1=O)CCCCCCCCCCC(=O)O
2
11-Maleimidoundecanoic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.