CAS RN 56961-86-5
CAS RN 56961-86-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
2,3,5-Trichlorotoluene, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
2
2,3,5-Trichlorotoluene, TRC
CAS: 56961-86-5 Molecular Formula: C7 H5 Cl3 Molecular Weight (g/mol): 195.474 Synonym: 1,2,5-Trichloro-3-methylbenzene (ACI),2,3,5-Trichlorotoluene,1,2,5-Trichloro-3-methylbenzene IUPAC Name: 1,2,5-trichloro-3-methylbenzene SMILES: Cc1cc(Cl)cc(Cl)c1Cl