accessibility menu, dialog, popup

UserName

CAS RN 56309-62-7

H3COOCH3NCO

CAS RN 56309-62-7

IUPAC Name: 2-isocyanato-1,4-dimethoxybenzene
Synonyms: 2-isocyanato-1,4-dimethoxybenzene
Molecular Weight (g/mol): 179.18
Molecular Formula: C9H9NO3
InChi Key: XNQBFHMCUNRKPM-UHFFFAOYSA-N
SMILES: COC1=CC(N=C=O)=C(OC)C=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)