accessibility menu, dialog, popup

UserName

CAS RN 562080-79-9

IUPAC Name: N-(4-bromophenyl)-3-(trifluoromethyl)benzamide
Synonyms: N-(4-bromophenyl)-3-(trifluoromethyl)benzamide
Molecular Weight (g/mol): 344.13
Molecular Formula: C14H9BrF3NO
InChi Key: JDUHFWWSDVISOZ-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=CC(=C1)C(=O)NC1=CC=C(Br)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results