accessibility menu, dialog, popup

UserName

CAS RN 56180-94-0

CH3(R)O(R)O(S)(R)OH(R)HO(R)O(R)OHO(S)(R)HO(R)HO(R)OHO(R)OH(R)OH(S)OH(S)NH(S)HO(S)OH(S)OH(S)OH

CAS RN 56180-94-0

IUPAC Name: (2R,3R,4R,5S,6R)-5-{[(2R,3R,4R,5S,6R)-5-{[(2R,3R,4S,5S,6R)-3,4-dihydroxy-6-methyl-5-{[(1S,4S,5S,6S)-4,5,6-trihydroxy-3-(hydroxymethyl)cyclohex-2-en-1-yl]amino}oxan-2-yl]oxy}-3,4-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-6-(hydroxymethyl)oxane-2,3,4-triol
Synonyms: acarbose bayer brand of acarbose acarbose (substance) acarbosum precose acarbose (product) glucobay glumida prandase acarbosa
Molecular Weight (g/mol): 645.61
Molecular Formula: C25H43NO18
InChi Key: XUFXOAAUWZOOIT-JMPDRRIHSA-N
SMILES: C[C@H]1O[C@H](O[C@@H]2[C@@H](CO)O[C@H](O[C@@H]3[C@@H](CO)O[C@@H](O)[C@H](O)[C@H]3O)[C@H](O)[C@H]2O)[C@H](O)[C@@H](O)[C@@H]1N[C@H]1C=C(CO)[C@H](O)[C@H](O)[C@H]1O
Molecular Weight (g/mol) 
  • (4)
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (5)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (4)
  • (3)
  • (2)
  • (3)
  • (5)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)