CAS RN 55804-65-4
CAS RN 55804-65-4
IUPAC Name:
4-oxo-3-oxa-13-azatetracyclo[7.7.1.0²,⁷.0¹³,¹⁷]heptadeca-1,5,7,9(17)-tetraene-5-carboxylate
Synonyms:
4-oxo-3-oxa-13-azatetracyclo[7.7.1.0²,⁷.0¹³,¹⁷]heptadeca-1,5,7,9(17)-tetraene-5-carboxylate
Molecular Weight (g/mol):
284.29
Molecular Formula:
C16H14NO4
InChi Key:
KCDCNGXPPGQERR-UHFFFAOYSA-M
SMILES:
[O-]C(=O)C1=CC2=CC3=C4N(CCC3)CCCC4=C2OC1=O
1
–
1
of
1
results
1
Promotions Available
Coumarin 343, pure, laser grade
Coumarin 343, pure, laser grade
CAS: 55804-65-4 Molecular Formula: C16H15NO4 Molecular Weight (g/mol): 285.299 MDL Number: MFCD00051335 InChI Key: KCDCNGXPPGQERR-UHFFFAOYSA-N PubChem CID: 108770 ChEBI: CHEBI:51941 SMILES: C1CC2=C3C(=C4C(=C2)C=C(C(=O)O4)C(=O)O)CCCN3C1