CAS RN 5571-36-8
CAS RN 5571-36-8
IUPAC Name:
11a-methyl-1,2,3,3a,3b,4,5,6,8,9,11,11a-dodecahydrospiro[cyclopenta[a]phenanthrene-7,2'-[1,3]dioxolan]-1-one
Synonyms:
11a-methyl-2,3,3a,3b,4,5,6,8,9,11-decahydrospiro[cyclopenta[a]phenanthrene-7,2'-[1,3]dioxolan]-1-one
Molecular Weight (g/mol):
314.43
Molecular Formula:
C20H26O3
InChi Key:
XUOQKQRMICQUQC-UHFFFAOYNA-N
SMILES:
CC12CC=C3C(CCC4=C3CCC3(C4)OCCO3)C1CCC2=O
1
–
3
of
3
results
1
Thermo Scientific Chemicals Estradiene dione
CAS: 5571-36-8 Molecular Formula: C20H26O3 Molecular Weight (g/mol): 314.43 InChI Key: XUOQKQRMICQUQC-UHFFFAOYNA-N IUPAC Name: 11a-methyl-1,2,3,3a,3b,4,5,6,8,9,11,11a-dodecahydrospiro[cyclopenta[a]phenanthrene-7,2'-[1,3]dioxolan]-1-one SMILES: CC12CC=C3C(CCC4=C3CCC3(C4)OCCO3)C1CCC2=O
2
Estra-5(10),9(11)-diene-3,17-dione 3-Ethylene Ketal, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Spiro[1,3-dioxolan-2,3′-estra-5′(10′),9′(11′)-dien]-17′-one, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.