CAS RN 55287-49-5
CAS RN 55287-49-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
Benzyl 2-Acetamido-2-deoxy-3,6-di-O-benzyl-Alpha-D-glucopyranoside, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Benzyl 2-Acetamido-2-deoxy-3,6-di-O-benzyl-Alpha-D-glucopyranoside, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Benzyl 2-Acetamido-2-deoxy-3,6-di-O-benzyl-Alpha-D-glucopyranoside, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.