CAS RN 55-27-6
CAS RN 55-27-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
DL-Norepinephrine Hydrochloride, TRC
CAS: 55-27-6 Molecular Formula: C8 H11 N O3 . Cl H Molecular Weight (g/mol): 205.64 Synonym: 1,2-Benzenediol, 4-(2-amino-1-hydroxyethyl)-, hydrochloride (1:1),1,2-Benzenediol, 4-(2-amino-1-hydroxyethyl)-, hydrochloride (9CI),1,2-Benzenediol, 4-(2-amino-1-hydroxyethyl)-, hydrochloride, (±)-,Benzyl alcohol, α-(aminomethyl)-3,4-dihydroxy-, hydrochloride, (±)- (8CI),(±)-Noradrenaline hydrochloride,(±)-Norepinephrine hydrochloride,DL-Arterenol hydrochloride,DL-Norepinephrine hydrochloride,NSC 7930,dl-Noradrenaline hydrochloride,dl-Norepinephrine hydrochloride IUPAC Name: 4-(2-amino-1-hydroxyethyl)benzene-1,2-diol;hydrochloride SMILES: Cl.NCC(O)c1ccc(O)c(O)c1